Northern Ireland
Order Entry
Anti-CD5 Mouse Monoclonal Antibody [clone: 8E7]
  ORIGCF802417
 :  OriGene
Anti-CD5 Mouse Monoclonal Antibody [clone: 8E7]
  ORIGCF802417
 :  OriGene
 
£781.00
   
  1 * 100 µl

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody type:
    Primary
  • Antigen symbol:
    CD5
  • Clonality:
    Monoclonal
  • Clone:
    8E7
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Epitope:
    Full length
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    54.4 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Full length human recombinant protein of human CD5 (NP_055022) produced in HEK293T cell.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µl

 

 

Recommended Dilutions: Flow Cytometry: 1:50

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 8E7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human