Northern Ireland
Order Entry
Anti-C17orf28 Mouse Monoclonal Antibody [clone: 4F6]
Anti-C17orf28 Mouse Monoclonal Antibody [clone: 4F6]
Catalog # ORIGCF501329
Supplier:  OriGene
Anti-C17orf28 Mouse Monoclonal Antibody [clone: 4F6]
Catalog # ORIGCF501329
Supplier:  OriGene
Price
£781.00
Log in for availability
Unit of Measure 1 * 100 µG

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody type:
    Primary
  • Antigen symbol:
    C17orf28
  • Clonality:
    Monoclonal
  • Clone:
    4F6
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    full length
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    88.6 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Full length human recombinant protein of human C17orf28 (NP_085133) produced in HEK293T cell.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

 

 

Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: C17orf28
Clonality: Monoclonal
Clone: 4F6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human