Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

You Searched For:

TLC Plate Applicators

4  results were found
https://ni.vwr.com/stibo/smallweb/std.lang.all/98/90/18989890.jpg

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"4"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  CAMAG
Description:   Disposable glass capillaries for manual sample application of 0,5, 1, 2, 5 or 10 µl. Handheld and can be positioned with the Multipurpose Spotting Gui...
Supplier:  CAMAG
Description:   Compact capillary dispensers can be used on the Nanomat or independently. The capillary pipettes are loaded into the dispenser in magazines from a pac...
Catalog Number: (552-0007)

Supplier:  CAMAG
Description:   The Nanomat 4 is used for easy application of samples in the form of spots onto TLC and HPTLC layers. A notch system ensures that sample positioning m...
UOM:  1 * 1 items
Catalog Number: (551-4230)

Supplier:  CAMAG
Description:   For plates up to 200×200 mm.
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is dicontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 4  of 4