Anti-Zinc Finger and Homeobox 1 Rabbit Polyclonal Antibody
Recommended dilution: ELISA:1:62500
Type: Primary
Antigen: Zinc Finger and Homeobox 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- USBIZ0469-60-100
- USBIZ0469-60-50
- Antigen name:
- Zinc Finger and Homeobox 1
- Zinc Finger and Homeobox 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Sequence:
- TDDSDTWEPPRHVKRKL
- Form:
- liquid
- liquid
- Purification:
- purified by immunoaffinity chromatography
- purified by immunoaffinity chromatography
- Concentration:
- ~0,1 mg/ml
- ~0,1 mg/ml
- Storage buffer:
- PBS, 0,02% sodium azide, before the addition of glycerol to 40%.
- PBS, 0,02% sodium azide, before the addition of glycerol to 40%.
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
- SizePk
Specifications
Antigen nameZinc Finger and Homeobox 1Environmentally PreferableAntibody typePrimaryClonalityPolyclonalConjugationUnconjugatedReactivityHumanHostRabbitIsotypeIgGELISAYesSequenceTDDSDTWEPPRHVKRKLFormliquidPurificationpurified by immunoaffinity chromatographyConcentration~0,1 mg/mlStorage bufferPBS, 0,02% sodium azide, before the addition of glycerol to 40%.Storage temperatureCoolShipping temperatureIceSpecifications
Antigen nameZinc Finger and Homeobox 1Environmentally PreferableAntibody typePrimaryClonalityPolyclonalConjugationUnconjugatedReactivityHumanHostRabbitIsotypeIgGELISAYesSequenceFormliquidPurificationpurified by immunoaffinity chromatographyConcentration~0,1 mg/mlStorage bufferPBS, 0,02% sodium azide, before the addition of glycerol to 40%.Storage temperatureCoolShipping temperatureIce