Anti-WISP3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Supplier: United States Biological
USBIW1450-53-APCEA 659 GBP
USBIW1450-53-APC
Anti-WISP3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:200; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: WISP3 (WNT1 inducible signaling pathway protein 3)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR