Anti-TFEB Goat Polyclonal Antibody

Supplier: United States Biological
USBIT8196-50FEA 532 GBP
USBIT8196-50F
Anti-TFEB Goat Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 0.5-2?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: TFEB (transcription factor EB)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR