Anti-SHBG Goat Polyclonal Antibody

Supplier: United States Biological
USBIS1012-52JEA 532 GBP
USBIS1012-52J
Anti-SHBG Goat Polyclonal Antibody
Antibodies
Recommended dilution: Immunochemistry: 3. 75 ?g/ml

Type: Primary
Antigen: SHBG (sex hormone-binding globulin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR