Anti-PPP1R2 Mouse Monoclonal Antibody [clone: 2E9]

Supplier: United States Biological
USBIP9107-30NEA 659 GBP
USBIP9107-30N
Anti-PPP1R2 Mouse Monoclonal Antibody [clone: 2E9]
Antibodies
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: PPP1R2 (protein phosphatase 1, regulatory (inhibitor) subunit 2)
Clonality: monoclonal
Clone: 2000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR