Anti-NPC1 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIN2566-31AEA 538 GBP
USBIN2566-31A
Anti-NPC1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NPC1 (Niemann-Pick disease, type C1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Hamster, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR