Anti-MMP15 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Supplier: United States Biological
USBIM2430-03C-PEEA 798 GBP
USBIM2430-03C-PE
Anti-MMP15 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Recommended dilution: Western Blot: 1:100-1:500; FLISA: 1:1000; Immunochemistry: 1:50-1:100; Flow Cytometry: 1:10-1:50
Do not freeze!

Type: Primary
Antigen: MMP15 (matrix metallopeptidase 15 (membrane-inserted))
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR