Anti-KLF16 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Supplier: United States Biological
USBIK1891-75A-APCEA 798 GBP
USBIK1891-75A-APC
Anti-KLF16 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: KLF16 (Kruppel-like factor 16)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR