Anti-HIPK2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: United States Biological
USBIH4065-10D-APEA 651 GBP
USBIH4065-10D-AP
Anti-HIPK2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: HIPK2 (homeodomain interacting protein kinase 2)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR