Anti-Estriol Mouse Monoclonal Antibody [clone: 9F12]

Supplier: United States Biological
USBIE3560-02EEA 541 GBP
USBIE3560-02E
Anti-Estriol Mouse Monoclonal Antibody [clone: 9F12]
Antibodies
Type: Primary
Antigen: Estriol
Clonality: monoclonal
Clone: 9F12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR