Anti-CD226 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 4j67]

Supplier: United States Biological
USBIC2548-97G4EA 513 GBP
USBIC2548-97G4
Anti-CD226 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 4j67]
Antibodies
Do not freeze!

Type: Primary
Antigen: CD226 (CD226 molecule)
Clonality: Monoclonal
Clone: 4j67
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR