Anti-CD4 Sheep Polyclonal Antibody

Supplier: United States Biological
USBIC2255-80JEA 728 GBP
USBIC2255-80J
Anti-CD4 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR