Anti-CSNK2A1 Rabbit polyclonal antibody

Supplier: United States Biological
USBIC2085-65PEA 532 GBP
USBIC2085-65P
Anti-CSNK2A1 Rabbit polyclonal antibody
Antibodies
Recommended dilution: Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: CSNK2A1 (casein kinase 2, alpha 1 polypeptide)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR