Anti-BCL2L11 Goat polyclonal antibody unconjugated

Supplier: United States Biological
USBIB1090-03GEA 516 GBP
USBIB1090-03G
Anti-BCL2L11 Goat polyclonal antibody unconjugated
Antibodies
Recommended dilution: ELISA: 1:16000; Western Blot: 0.5-2?g/ml

Type: Primary
Antigen: BCL2L11 (BCL2-like 11 (apoptosis facilitator))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR