Anti-APOE Goat polyclonal antibody unconjugated

Supplier: United States Biological
USBIA2299-74JEA 252 GBP
USBIA2299-74J
Anti-APOE Goat polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: APOE (apolipoprotein E)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR