Anti-APBB1 Goat Polyclonal Antibody

Supplier: United States Biological
USBIA2275-71QEA 701 GBP
USBIA2275-71Q
Anti-APBB1 Goat Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:128000; Western Blot: 0.1-0. 3?g/ml

Type: Primary
Antigen: APBB1 (amyloid beta (A4) precursor protein-binding, family B, member 1 (Fe65))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR