Anti-Rabbit Serum Albumin Goat Polyclonal Antibody
USBIA1327-70E
- Antibody type:Primary
- Antigen symbol:Rabbit Serum Albumin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Rabbit
- Environmentally Preferable:
- Format:Serum
- Form:lyophilized powder
- Storage buffer:sterile ddH2O
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Type: Primary
Antigen: Rabbit Serum Albumin (Rabbit Serum Albumin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit