Anti-ACTB Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIA0760-41REA 573 GBP
USBIA0760-41R
Anti-ACTB Rabbit Polyclonal Antibody
Antibodies
  • Recommended dilution: ELISA: 1:5000-1:20000; Immunochemistry: 1:50-1:200; Western Blot: 1:500-1:1000

Type: Primary
Antigen: ACTB (actin, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR