Anti-ZNF217 Goat Polyclonal Antibody

Supplier: United States Biological
USBI151171EA 701 GBP
USBI151171
Anti-ZNF217 Goat Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:16000

Type: Primary
Antigen: ZNF217 (zinc finger protein 217)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR