Anti-TRAF1 Goat Polyclonal Antibody

Supplier: United States Biological
USBI151091EA 598 GBP
USBI151091
Anti-TRAF1 Goat Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:16000; Western Blot: 0. 3-1 ?g/ml

Type: Primary
Antigen: TRAF1 (TNF receptor-associated factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR