Anti-SH2D1A Goat Polyclonal Antibody

Supplier: United States Biological
USBI150967EA 701 GBP
USBI150967
Anti-SH2D1A Goat Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:16000; Western Blot: 0. 3-1 ?g/ml

Type: Primary
Antigen: SH2D1A (SH2 domain containing 1A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR