Anti-PTPRT Goat Polyclonal Antibody

Supplier: United States Biological
USBI150870EA 598 GBP
USBI150870
Anti-PTPRT Goat Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:32000; Western Blot: 0. 3-1 ?g/ml

Type: Primary
Antigen: PTPRT (protein tyrosine phosphatase, receptor type, T)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR