Anti-UBE2R2 Mouse Monoclonal Antibody [clone: 5E7]

Supplier: United States Biological
USBI135013EA 489 GBP
USBI135013
Anti-UBE2R2 Mouse Monoclonal Antibody [clone: 5E7]
Antibodies
Recommended dilution: ELISA: 0.1ng/ml

Type: Primary
Antigen: UBE2R2 (ubiquitin-conjugating enzyme E2R 2)
Clonality: monoclonal
Clone: 50000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR