Anti-SOCS5 Mouse Polyclonal Antibody

Supplier: United States Biological
USBI133688EA 489 GBP
USBI133688
Anti-SOCS5 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SOCS5 (suppressor of cytokine signaling 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR