Anti-PKD1L2 Rabbit Polyclonal Antibody

Supplier: US Biological
USBI131386EA 464 GBP
USBI131386
Anti-PKD1L2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PKD1L2 (polycystic kidney disease 1-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR