Anti-OTUB2 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBI130770EA 464 GBP
USBI130770
Anti-OTUB2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: OTUB2 (OTU deubiquitinase, ubiquitin aldehyde binding 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR