Anti-KLRAP1 Mouse Monoclonal Antibody [clone: 1H3]

Supplier: United States Biological
USBI128897EA 435 GBP
USBI128897
Anti-KLRAP1 Mouse Monoclonal Antibody [clone: 1H3]
Antibodies
Type: Primary
Antigen: KLRAP1 (killer cell lectin-like receptor subfamily A pseudogene 1)
Clonality: monoclonal
Clone: 1H3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR