Anti-CROT Mouse Monoclonal Antibody [clone: 1A6]

Supplier: United States Biological
USBI125350EA 489 GBP
USBI125350
Anti-CROT Mouse Monoclonal Antibody [clone: 1A6]
Antibodies
Type: Primary
Antigen: CROT (carnitine O-octanoyltransferase)
Clonality: monoclonal
Clone: 1A6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR