Anti-GAPR1 Mouse Polyclonal Antibody

Supplier: US Biological
USBI124167EA 489 GBP
USBI124167
Anti-GAPR1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GAPR1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR