Anti-ZNF398 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBI044221EA 550 GBP
USBI044221
Anti-ZNF398 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: ZNF398 (zinc finger protein 398)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR