Anti-ZEB2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: United States Biological
USBI044055-APEA 798 GBP
USBI044055-AP
Anti-ZEB2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: ZEB2 (zinc finger E-box binding homeobox 2)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR