Anti-FIGF Rabbit Polyclonal Antibody

Supplier: United States Biological
USBI043716EA 550 GBP
USBI043716
Anti-FIGF Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:250-500; Immunochemistry: 1:50-100

Type: Primary
Antigen: FIGF (c-fos induced growth factor (vascular endothelial growth factor D))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR