Anti-TRA2A Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: United States Biological
USBI043176-APEA 651 GBP
USBI043176-AP
Anti-TRA2A Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: TRA2A (transformer 2 alpha homolog (Drosophila))
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR