Anti-PTP4A1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: United States Biological
USBI040650-FITCEA 651 GBP
USBI040650-FITC
Anti-PTP4A1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: PTP4A1 (protein tyrosine phosphatase type IVA, member 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR