Anti-PSMG2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Supplier: United States Biological
USBI040572-APCEA 651 GBP
USBI040572-APC
Anti-PSMG2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: PSMG2 (proteasome (prosome, macropain) assembly chaperone 2)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR