Anti-PRRX2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: United States Biological
USBI040516-HRPEA 798 GBP
USBI040516-HRP
Anti-PRRX2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: PRRX2 (paired related homeobox 2)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR