Anti-INPP5B Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Supplier: US Biological
USBI037054-APCEA 651 GBP
USBI037054-APC
Anti-INPP5B Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: INPP5B (inositol polyphosphate-5-phosphatase, 75kDa)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR