Anti-CLEC12B Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Supplier: US Biological
USBI033975-APCEA 798 GBP
USBI033975-APC
Anti-CLEC12B Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!

Type: Primary
Antigen: CLEC12B (C-type lectin domain family 12, member B)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR