Anti-APCDD1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Supplier: US Biological
USBI031956-PEEA 798 GBP
USBI031956-PE
Anti-APCDD1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: APCDD1 (adenomatosis polyposis coli down-regulated 1)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR