Anti-IgM µ Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
ROCK612-4107EA 269 GBP
ROCK612-4107
Anti-IgM µ Rabbit Polyclonal Antibody
Antibodies
Secondary Rabbit Anti-IgM (mu chain) Reacts with Rat

Recommended Dilutions: ELISA: 1:50000 - 1:250000; Western Blot: 1:5000 - 1:25000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: µ-chain
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR