Anti-HIF3A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
ROCK600-401-435EA 734 GBP
ROCK600-401-435
Anti-HIF3A Rabbit Polyclonal Antibody
Antibodies
This affinity-purified antibody has been tested for use in ELISA and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 72 kDa in size corresponding to Hif3 a Protein by western blotting.

Recommended Dilutions: ELISA: 1:14000 - 1:80,000; Western Blot: 1:1000 - 1:8000;

Type: Primary
Antigen: HIF3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR