Anti-IgG (H+L) Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
ROCK307-4102EA 413 GBP
ROCK307-4102
Anti-IgG (H+L) Rabbit Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Golden Syrian Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR