Anti-IgG F(ab')2 Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
ROCK210-1104EA 458 GBP
ROCK210-1104
Anti-IgG F(ab')2 Goat Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR