Anti-TRIM2 Goat Polyclonal Antibody

Supplier: Everest Biotechnology
EVBIEB10871EA 317 GBP
EVBIEB10871
Anti-TRIM2 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TRIM2 (tripartite motif containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR