Anti-ALDH5A1 Goat Polyclonal Antibody

Supplier: Everest Biotechnology
EVBIEB10436EA 317 GBP
EVBIEB10436
Anti-ALDH5A1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ALDH5A1 (aldehyde dehydrogenase 5 family, member A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR