Anti-NRAP Goat Polyclonal Antibody

Supplier: Everest Biotechnology
EVBIEB10399EA 189 GBP
EVBIEB10399
Anti-NRAP Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NRAP (nebulin-related anchoring protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR