Anti-MTHFD2L Goat Polyclonal Antibody

Supplier: Everest Biotechnology
EVBIEB08463EA 317 GBP
EVBIEB08463
Anti-MTHFD2L Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MTHFD2L (methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR